Dimethylbis(t-butylcyclopentadienyl)zirconium ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

40-1110-5G 40-1110-1G
100199-872EA 867.63 USD
100199-872 100200-268
Dimethylbis(t-butylcyclopentadienyl)zirconium ≥98%
Dimethylbis(t-butylcyclopentadienyl)zirconium

Metallocenes, Derivatives & Cp Precursors, Volatile Organometallics for CVD & ALD, Volatile Precursors for CVD

MDL Number: MFCD01073787
CAS Number: 68193-40-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR