Cyclopentadienylzirconium trichloride ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

40-1100-5G 40-1100-1G
100198-752EA 341.11 USD
100198-752 100198-750
Cyclopentadienylzirconium trichloride ≥98%
Cyclopentadienylzirconium trichloride

Metallocenes, Derivatives & Cp Precursors

Formula: C₅H₅Cl₃Zr
Melting Pt: 237 °C (dec.)
MDL Number: MFCD00015756
CAS Number: 34767-44-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR