n-Butylcyclopentadienylzirconium trichloride

Supplier: STREM CHEMICALS INC MS
Warning

40-1080-1G 40-1080-5G
100207-396EA 244.86 USD
100207-396 100196-946
n-Butylcyclopentadienylzirconium trichloride
n-Butylcyclopentadienylzirconium trichloride

Metallocenes, Derivatives & Cp Precursors

MDL Number: MFCD02684560
CAS Number: 329735-75-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR