Bis(tetramethylcyclopentadienyl)zirconium(IV) dichloride ≥97%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
40-1060-25G
40-1060-1G
40-1060-5G
100207-394EA
1774.89
USD
100207-394
100200-948
100199-236
Bis(tetramethylcyclopentadienyl)zirconium(IV) dichloride ≥97%
Bis(tetramethylcyclopentadienyl)zirconium(IV) dichloride
Formula:
C₁₈H₂₆Cl₂Zr |
MDL Number:
MFCD00672114 CAS Number: 119445-90-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...