Bis(pentamethylcyclopentadienyl)zirconium(IV) dichloride ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Bis(pentamethylcyclopentadienyl)zirconium dichloride

40-1044-5G 40-1044-1G
100200-944EA 645.42 USD
100200-944 100200-942
Bis(pentamethylcyclopentadienyl)zirconium(IV) dichloride ≥99%
Bis(pentamethylcyclopentadienyl)zirconium(IV) dichloride

Metallocenes, Derivatives & Cp Precursors

Formula: C₂₀H₃₀Cl₂Zr
MW: 432.582 g/mol
MDL Number: MFCD00058849
CAS Number: 54039-38-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR