Bis(2-methylindenyl)zirconium dichloride ≥98%

Supplier: STREM CHEMICALS INC MS
Danger

40-1042-5G 40-1042-1G
100208-026EA 958.22 USD
100208-026 100207-392
Bis(2-methylindenyl)zirconium dichloride ≥98%
Bis(2-methylindenyl)zirconium dichloride

Metallocenes, Derivatives & Cp Precursors

MDL Number: MFCD00672121
CAS Number: 165688-64-2
UN: 3261
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR