Bis(indenyl)zirconium dichloride ≥98%

Supplier: STREM CHEMICALS INC MS
Danger

40-1041-1G 40-1041-5G 40-1041-25G
100199-868EA 131.63 USD
100199-868 100199-870 100200-940
Bis(indenyl)zirconium dichloride ≥98%
Bis(indenyl)zirconium dichloride

Metallocenes, Derivatives & Cp Precursors

MDL Number: MFCD00672104
CAS Number: 12148-49-1
UN: 3261
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR