Bis(cyclopentadienyl)zirconium hydridochloride ≥95%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Bis(cyclopentadienyl)zirconium(IV) chloride hydride, Schwartz's Reagent, Bis(cyclopentadienyl)zirconium chloride hydride

40-1040-1G 40-1040-5G 40-1040-25G
100207-664EA 56.62 USD
100207-664 100208-890 100200-678
Bis(cyclopentadienyl)zirconium hydridochloride ≥95%
Bis(cyclopentadienyl)zirconium hydridochloride

Metallocenes, Derivatives & Cp Precursors

Formula: C₁₀H₁₁ClZr
MW: 257.87 g/mol
Melting Pt: > 300 °C
MDL Number: MFCD02089401
CAS Number: 37342-97-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR