Bis(ethylcyclopentadienyl)zirconium dichloride ≥97%

Supplier: STREM CHEMICALS INC MS
Warning

40-1030-25G 40-1030-5G 40-1030-1G
100203-112EA 3300.68 USD
100203-112 100203-114 100209-342
Bis(ethylcyclopentadienyl)zirconium dichloride ≥97%
Bis(ethylcyclopentadienyl)zirconium dichloride

Metallocenes, Derivatives & Cp Precursors

MDL Number: MFCD00269854
CAS Number: 73364-08-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR