1,1'-Dibutylzirconocene dichloride ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Bis(n-butylcyclopentadienyl)zirconium dichloride, Bis(butylcyclopentadienyl)zirconium(IV) Dichloride

40-0900-1G 40-0900-5G
100209-916EA 133.05 USD
100209-916 100209-918
1,1'-Dibutylzirconocene dichloride ≥98%
1,1'-Dibutylzirconocene dichloride

Metallocenes, Derivatives & Cp Precursors

Formula: C₁₈H₂₆Cl₂Zr
MW: 404.53 g/mol
MDL Number: MFCD00672105
CAS Number: 73364-10-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR