Bis(tert-butylcyclopentadienyl)zirconium(IV) dichloride ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Bis(t-butylcyclopentadienyl)zirconium dichloride

40-0750-1G 40-0750-5G
100200-938EA 169.85 USD
100200-938 100201-212
Bis(tert-butylcyclopentadienyl)zirconium(IV) dichloride ≥98%
Bis(tert-butylcyclopentadienyl)zirconium(IV) dichloride

Metallocenes, Derivatives & Cp Precursors

Formula: C₁₈H₂₆Cl₂Zr
MW: 404.529 g/mol
MDL Number: MFCD01073785
CAS Number: 32876-92-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR