Yttrium(III) neodecanoate ≥99.9% (Y basis) (12-16% Y), Superconductor Grade

Supplier: STREM CHEMICALS INC MS

39-7000-50G 39-7000-10G 39-7000-250G
100202-764EA 318.46 USD
100202-764 100205-110 100207-390
Yttrium(III) neodecanoate ≥99.9% (Y basis) (12-16% Y), Superconductor Grade
Yttrium(III) neodecanoate
Formula: Y(OOC₁₀H₁₉)₃
MDL Number: MFCD00070443
CAS Number: 68683-17-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR