Tris(n-propylcyclopentadienyl)yttrium ≥99.9%

Supplier: STREM CHEMICALS INC MS
Danger

39-5100-5G 39-5100-1G
100202-670EA 912.92 USD
100202-670 100208-420
Tris(n-propylcyclopentadienyl)yttrium ≥99.9%
Tris(n-propylcyclopentadienyl)yttrium

Metallocenes, Derivatives & Cp Precursors, Volatile Organometallics for CVD & ALD, Volatile Precursors for CVD

MDL Number: MFCD00145488
CAS Number: 329735-73-1
UN: 1325
ADR: 4.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR