Tris(methylcyclopentadienyl)yttrium(III) ≥99.9%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Tris(methylcyclopentadienyl)yttrium

39-5050-1G 39-5050-5G
100208-418EA 189.66 USD
100208-418 100202-668
Tris(methylcyclopentadienyl)yttrium(III) ≥99.9%
Tris(methylcyclopentadienyl)yttrium(III)

Metallocenes, Derivatives & Cp Precursors, Volatile Organometallics for CVD & ALD, Volatile Precursors for CVD

Formula: C₁₈H₂₁Y
MW: 326.272 g/mol
MDL Number: MFCD00145485
CAS Number: 329735-72-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR