Tris(cyclopentadienyl)yttrium(III) ≥99.9%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Tris(cyclopentadienyl)yttrium

39-5000-1G 39-5000-5G
100203-774EA 182.58 USD
100203-774 100202-400
Tris(cyclopentadienyl)yttrium(III) ≥99.9%
Tris(cyclopentadienyl)yttrium(III)

Metallocenes, Derivatives & Cp Precursors, Volatile Organometallics for CVD & ALD, Volatile Precursors for CVD

Formula: C₁₅H₁₅Y
MW: 284.19 g/mol
Boiling Pt: 200 °C (subl. in vacuum)
Melting Pt: 295 °C
MDL Number: MFCD00058706
CAS Number: 1294-07-1
UN: 1325
ADR: 4.1 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR