Tris(n-butylcyclopentadienyl)yttrium(III) ≥99.9%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Tris(butylcyclopentadienyl)yttrium

39-4950-1G 39-4950-5G
100208-416EA 154.28 USD
100208-416 100202-666
Tris(n-butylcyclopentadienyl)yttrium(III) ≥99.9%
Tris(n-butylcyclopentadienyl)yttrium(III)

Metallocenes, Derivatives & Cp Precursors, Volatile Organometallics for CVD & ALD, Volatile Precursors for CVD

Formula: C₂₇H₃₉Y
MW: 452.5 g/mol
MDL Number: MFCD00145473
CAS Number: 312739-77-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR