Yttrium(III) trifluoromethanesulphonate ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Yttrium(III) trifluoromethanesulphonate, Yttrium triflate

39-4050-25G 39-4050-5G
101442-490EA 376.49 USD
101442-490 101442-492
Yttrium(III) trifluoromethanesulphonate ≥98%
Yttrium(III) trifluoromethanesulphonate

Metal Triflate

Formula: C₃F₉O₉S₃Y
MW: 536.12 g/mol
MDL Number: MFCD00209623
CAS Number: 52093-30-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR