Yttrium(III) phosphate hydrate ≥99.99%, PURATREM

Supplier: STREM CHEMICALS INC MS

39-2980-5G 39-2980-25G
100197-504EA 138.71 USD
100197-504 100197-502
Yttrium(III) phosphate hydrate ≥99.99%, PURATREM
Yttrium(III) phosphate hydrate
Formula: YPO₄·xH₂O
MW: 183.88 g/mol (anhy)
MDL Number: MFCD00049782
CAS Number: 34054-55-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR