Tris[N,N-bis(trimethylsilyl)amide]yttrium(III) ≥98% (99.9%)

Supplier: STREM CHEMICALS INC MS
Warning

39-1500-5G 39-1500-1G 39-1500-25G
100206-596EA 413.29 USD
100206-596 100200-932 100209-256
Tris[N,N-bis(trimethylsilyl)amide]yttrium(III) ≥98% (99.9%)
Tris[N,N-bis(trimethylsilyl)amide]yttrium (III)

Metal Alkylamides, Volatile Precursors for CVD

Formula: C₁₈H₅₄N₃Si₆Y
Boiling Pt: 105 °C (10…4 mmHg) (subl.)
Melting Pt: 180…184 °C
MDL Number: MFCD00210649
CAS Number: 41836-28-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR