Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')yttrium(III) ≥98% (99.9%-Y)
Supplier: STREM CHEMICALS INC MS
Synonyms:
Y(TMHD)3, Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)yttrium(III)
39-1000-25G
39-1000-1G
39-1000-5G
100206-592EA
1317.72
USD
100206-592
100204-044
100206-594
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')yttrium(III) ≥98% (99.9%-Y)
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')yttrium(III)
Metal Beta-diketonates, Metal TMHD, Volatile Precursors for CVD
Formula:
C₃₃H₅₇O₆Y MW: 638.708 g/mol Melting Pt: 170…173 °C |
MDL Number:
MFCD00015713 CAS Number: 15632-39-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...