Strontium hexafluoroacetylacetonate

Supplier: STREM CHEMICALS INC MS
Warning

38-2000-25G 38-2000-5G 38-2000-1G
100201-208EA 870.46 USD
100201-208 100208-022 100205-104
Strontium hexafluoroacetylacetonate
Strontium hexafluoroacetylacetonate

Metal Beta-diketonates, Volatile Precursors for CVD

Boiling Pt: 220 °C (0.02 mmHg) (subl.)
Melting Pt: 260 °C (dec.)
MDL Number: MFCD00064760
CAS Number: 1245785-21-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR