Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)strontium tetraglyme adduct ≥99.99%, PURATREM

Supplier: STREM CHEMICALS INC MS

38-1010-1G 38-1010-5G
100201-602EA 181.17 USD
100201-602 100205-934
Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)strontium tetraglyme adduct ≥99.99%, PURATREM
Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)strontium tetraglyme adduct

Metal Beta-diketonates, Metal TMHD, Volatile Precursors for CVD

Melting Pt: 75 °C
MDL Number: MFCD01073782
CAS Number: 150939-76-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR