Bis(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')strontium hydrate
Supplier: STREM CHEMICALS INC MS
Synonyms:
Sr(TMHD)2, Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)strontium hydrate
38-1000-5G
38-1000-1G
38-1000-25G
100199-862EA
339.69
USD
100199-862
100205-102
100199-860
Bis(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')strontium hydrate
Bis(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')strontium hydrate
Metal Beta-diketonates, Metal TMHD, Volatile Precursors for CVD
Formula:
C₂₂H₃₈O₄Sr·H₂O Boiling Pt: 250 °C/ 230 °C (0.05 mmHg) (dec./ subl.) Melting Pt: 200…203 °C |
MDL Number:
MFCD00151017 CAS Number: 199445-30-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...