Selenourea ≥99%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
34-1900-25G
34-1900-5G
100211-070EA
1561.17
USD
100211-070
100211-072
Selenourea ≥99%
Selenourea
Formula:
CH₄N₂Se Melting Pt: 200 °C (dec.) |
MDL Number:
MFCD00008065 CAS Number: 630-10-4 UN: 3283 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...