Zinc 2-ethylhexanoate ≥99% (~18% Zn, 1% Diethylene glycol monomethyl ether)
Supplier: STREM CHEMICALS INC MS
Synonyms:
Zinc(II) 2-ethylhexanoate
30-3038-500G
30-3038-100G
100200-924EA
144.37
USD
100200-924
100209-902
Zinc 2-ethylhexanoate ≥99% (~18% Zn, 1% Diethylene glycol monomethyl ether)
Zinc 2-ethylhexanoate
Formula:
C₁₆H₃₀O₄Zn MW: 351.792 g/mol Flash Pt: 260 °F |
MDL Number:
MFCD00053316 CAS Number: 136-53-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...