Zinc (II) phthalocyanine (Sulfate ionophore II) ≥95%
Supplier: STREM CHEMICALS INC MS
Synonyms:
Zinc phthalocyanine, 29H,31H-Phthalocyanine, zinc complex
30-3000-10G
30-3000-2G
100200-456EA
186.83
USD
100200-456
100198-742
Zinc (II) phthalocyanine (Sulfate ionophore II) ≥95%
Zinc (II) phthalocyanine (Sulphate ionophore II)
Formula:
C₃₂H₁₆N₈Zn MW: 577.91 g/mol |
MDL Number:
MFCD00041955 CAS Number: 14320-04-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...