Copper(II) neodecanoate ∼60% in toluene (6-12% Cu), Superconductor Grade

Supplier: STREM CHEMICALS INC MS
Danger

29-7000-50G 29-7000-10G
100201-590EA 608.62 USD
100201-590 100204-028
Copper(II) neodecanoate ∼60% in toluene (6-12% Cu), Superconductor Grade
Copper(II) neodecanoate
MDL Number: MFCD00070437
CAS Number: 50315-14-5
UN: 1993
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR