Cupric naphthenate ≥77% in mineral spirits (8% Cu)

Supplier: STREM CHEMICALS INC MS
Danger

29-6750-250G 29-6750-1000G
100196-192EA 65.11 USD
100196-192 100205-092
Cupric naphthenate ≥77% in mineral spirits (8% Cu)
Cupric naphthenate
Formula: Cu(C₁₁H₇O₂)₂
Flash Pt: 104 °F
MDL Number: MFCD00146564
CAS Number: 1338-02-9
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR