Cyclopentadienyl(triethylphosphine)copper(I) ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

29-5500-1G 29-5500-5G
100200-266EA 172.68 USD
100200-266 100196-878
Cyclopentadienyl(triethylphosphine)copper(I) ≥98%
Cyclopentadienyl(triethylphosphine)copper(I)

Metallocenes, Derivatives & Cp Precursors, Volatile Organometallics for CVD & ALD, Volatile Precursors for CVD

Boiling Pt: 60 °C (0.01 mmHg) (subl.)
MDL Number: MFCD01074447
CAS Number: 12261-30-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR