Chloro[1,3-bis(2,6-diisopropylphenyl)imidazol-2-ylidene]copper(I) ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Chloro[1,3-bis(2,6-di-i-propylphenyl)imidazol-2-ylidene]copper(I), [1,3-Bis(2,6-diisopropylphenyl)imidazol-2-ylidene]copper chloride

29-4050-500MG 29-4050-2G
101418-802EA 148.62 USD
101418-802 101418-804
Chloro[1,3-bis(2,6-diisopropylphenyl)imidazol-2-ylidene]copper(I) ≥98%
Chloro[1,3-bis(2,6-diisopropylphenyl)imidazol-2-ylidene]copper(I)

NHC

Formula: C₂₇H₃₆ClCuN₂
MW: 487.60 g/mol
MDL Number: MFCD09264276
CAS Number: 578743-87-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR