Bromo(1,10-phenanthroline)(triphenylphosphine)copper(I) ≥97%

Supplier: STREM CHEMICALS INC MS
Warning

29-4000-1G 29-4000-5G
101324-678EA 110.4 USD
101324-678 101324-680
Bromo(1,10-phenanthroline)(triphenylphosphine)copper(I) ≥97%
Bromo(1,10-phenanthroline)(triphenylphosphine)copper(I)
Formula: C₃₀H₂₄BrCuN₂P
MW: 586.9487 g/mol
MDL Number: MFCD07369032
CAS Number: 25753-84-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR