Bis(triphenylphosphine)copper(I) nitrate ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

29-3100-25G 29-3100-5G
101324-674EA 312.8 USD
101324-674 101324-676
Bis(triphenylphosphine)copper(I) nitrate ≥99%
Bis(triphenylphosphine)copper(I) nitrate
MDL Number: MFCD02091690
CAS Number: 106678-35-7
UN: 1479
ADR: 5.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR