Bis(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')copper(II) ≥99%

Supplier: STREM CHEMICALS INC MS

Synonyms: Cu(TMHD)2, Copper(II)bis(2,2,6,6-tetramethyl-3,5-heptanedionate), Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)copper(II)

29-3000-25G 29-3000-1G 29-3000-5G
100197-484EA 854.89 USD
100197-484 100197-482 100197-486
Bis(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')copper(II) ≥99%
Bis(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')copper(II)

Metal Beta-diketonates, Metal TMHD, Volatile Precursors for CVD

Formula: C₂₂H₃₈CuO₄
MW: 430.08 g/mol
Boiling Pt: 315 °C/ 88 °C (0.05 mmHg) (dec./ subl.)
Melting Pt: 198 °C
MDL Number: MFCD00058920
CAS Number: 14040-05-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR