Copper(II) hexafluoroacetylacetonate hydrate ≥99.99% (Cu basis), PURATREM, Electronic Grade

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Hexafluoroacetylacetono Copper(II) Salt, Copper(II) Hexafluoroacetylacetonate, Copper(II) hexafluoro-2,4-pentanedionate hydrate, Bis(hexafluoroacetylacetonato)copper(II) hydrate, Copper(II) hexafluoro-2,4-pentanedionate xhydrate

29-2929-1G 29-2929-5G 29-2929-25G
100201-582EA 66.52 USD
100201-582 100211-064 100211-062
Copper(II) hexafluoroacetylacetonate hydrate ≥99.99% (Cu basis), PURATREM, Electronic Grade
Copper (II) hexafluoroacetylacetonate hydrate

Metal Beta-diketonates, Volatile Precursors for CVD

Formula: C₁₀H₂CuF₁₂O₄·xH₂O
MW: 477.64 g/mol (anhy)
Boiling Pt: 220 °C/ 70 °C (0.05 mmHg) (dec./ subl.)
Melting Pt: 85…89 °C
MDL Number: MFCD00151019
CAS Number: 155640-85-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR