Nickel(II) meso-tetraphenylporphine

Supplier: STREM CHEMICALS INC MS

Synonyms: 5,10,15,20-Tetraphenylporphyrin nickel, 5,10,15,20-Tetraphenyl-21H,23H-porphine nickel(II), Nickel TPP

28-1690-5G 28-1690-1G
100203-764EA 1288 USD
100203-764 100208-014
Nickel(II) meso-tetraphenylporphine
Nickel(II) meso-tetraphenylporphine

Porphine

Formula: C₄₄H₂₈N₄Ni
MW: 671.41 g/mol
MDL Number: MFCD00010727
CAS Number: 14172-92-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR