Nickel(II) phthalocyanine ≥94%

Supplier: STREM CHEMICALS INC MS

28-1600-5G 28-1600-25G
100205-076EA 138.71 USD
100205-076 100201-200
Nickel(II) phthalocyanine ≥94%
Nickel(II) phthalocyanine

Volatile Precursors for CVD

Formula: Ni(C₃₂H₁₆N₈)
MW: 571.22 g/mol
MDL Number: MFCD00041954
CAS Number: 14055-02-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR