Nickel(II) oxalate dihydrate ≥99.998% (Ni basis), PURATREM

Supplier: STREM CHEMICALS INC MS

Synonyms: Nickel oxalate dihydrate

28-1470-50G 28-1470-10G
100207-380EA 644 USD
100207-380 100199-376
Nickel(II) oxalate dihydrate ≥99.998% (Ni basis), PURATREM
Nickel(II) oxalate dihydrate
Formula: C₂H₄NiO₆
MW: 182.741 g/mol
MDL Number: MFCD00167155
CAS Number: 6018-94-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR