Bis(cyclopentadienyl)nickel ≥99%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Nickelocene

28-1301-100G 28-1301-5G 28-1301-25G
100198-734EA 840.74 USD
100198-734 100209-886 100209-884
Bis(cyclopentadienyl)nickel ≥99%
Bis(cyclopentadienyl)nickel

Metallocenes, Derivatives & Cp Precursors, Volatile Metal Carbonyls for CVD & ALD, Volatile Organometallics for CVD & ALD, Volatile Precursors for CVD

Formula: C₁₀H₁₀Ni
Melting Pt: 173…174 °C
MDL Number: MFCD00001441
CAS Number: 1271-28-9
UN: 1325
ADR: 4.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR