Nickel acetate tetrahydrate ≥99.999% (Ni basis), PURATREM

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Nickel diacetate tetrahydrate, Nickel (II) acetate tetrahydrate, Nickel di(acetate) tetrahydrate, Acetic acid nickel (II) salt tetrahydrate

28-1100-25G 28-1100-5G
100211-054EA 400.55 USD
100211-054 100211-056
Nickel acetate tetrahydrate ≥99.999% (Ni basis), PURATREM
Nickel acetate tetrahydrate
Formula: C₄H₆NiO₄·4H₂O
MW: 248.84 g/mol
Melting Pt: Dec.
Density: 1.744
MDL Number: MFCD00066973
CAS Number: 6018-89-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR