Bis(triphenylphosphine)nickel(II) chloride ≥99%

Supplier: STREM CHEMICALS INC MS

Synonyms: Dichlorobis(triphenylphosphine)nickel(II), Bis(triphenylphosphine)nickel(II)chloride

28-0095-25G 28-0095-5G
100202-658EA 127.38 USD
100202-658 100201-568
Bis(triphenylphosphine)nickel(II) chloride ≥99%
Bis(triphenylphosphine)nickel(II) chloride
Formula: C₃₆H₃₂Cl₂NiP₂
MW: 656.19 g/mol
MDL Number: MFCD00009592
CAS Number: 14264-16-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR