Bis(triphenylphosphine)nickel(II) bromide ≥99%

Supplier: STREM CHEMICALS INC MS

Synonyms: Nickel(II) bromide bis(triphenylphosphine), Dibromobis(triphenylphosphine)nickel(II)

28-0092-50G 28-0092-10G
100201-566EA 396.31 USD
100201-566 100208-406
Bis(triphenylphosphine)nickel(II) bromide ≥99%
Bis(triphenylphosphine)nickel(II) bromide
Formula: C₃₆H₃₀Br₂NiP₂
MW: 743.08
Melting Pt: 219…223 °C
MDL Number: MFCD00015865
CAS Number: 14126-37-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR