Bis(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')nickel(II) ≥98% (99.9%-Ni)
Supplier: STREM CHEMICALS INC MS
Synonyms:
Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)nickel(II), Ni(TMHD)2
28-0088-5G
28-0088-1G
28-0088-25G
100205-072EA
266.09
USD
100205-072
100205-068
100205-070
Bis(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')nickel(II) ≥98% (99.9%-Ni)
Bis(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')nickel(II)
Metal Beta-diketonates, Metal TMHD, Volatile Precursors for CVD
Formula:
C₂₂H₃₈NiO₄ MW: 425.23 g/mol Boiling Pt: 90…110 °C (0,1 mmHg) (subl.) Melting Pt: 223…225 °C |
MDL Number:
MFCD00192348 CAS Number: 14481-08-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...