Bis(tetramethylcyclopentadienyl)nickel(II) ≥98%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Bis(tetramethylcyclopentadienyl)nickel
28-0087-5G
28-0087-1G
100198-730EA
1146.46
USD
100198-730
100208-012
Bis(tetramethylcyclopentadienyl)nickel(II) ≥98%
Bis(tetramethylcyclopentadienyl)nickel (II)
Metallocenes, Derivatives & Cp Precursors
Formula:
C₁₈H₂₆Ni |
MDL Number:
MFCD01862460 CAS Number: 79019-60-6 UN: 1325 ADR: 4.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...