Bis(i-propylcyclopentadienyl)nickel ≥98%

Supplier: STREM CHEMICALS INC MS
Danger

28-0086-5G 28-0086-1G
100202-396EA 734.58 USD
100202-396 100205-066
Bis(i-propylcyclopentadienyl)nickel ≥98%
Bis(i-propylcyclopentadienyl)nickel

Metallocenes, Derivatives & Cp Precursors, Volatile Organometallics for CVD & ALD, Volatile Precursors for CVD

MDL Number: MFCD01862459
CAS Number: 57197-55-4
UN: 1993
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR