Bis(pentamethylcyclopentadienyl)nickel(II) ≥98%

Supplier: STREM CHEMICALS INC MS
Danger

28-0085-5G 28-0085-1G
100200-450EA 768.55 USD
100200-450 100200-448
Bis(pentamethylcyclopentadienyl)nickel(II) ≥98%
Bis(pentamethylcyclopentadienyl)nickel (II)

Metallocenes, Derivatives & Cp Precursors, Volatile Organometallics for CVD & ALD, Volatile Precursors for CVD

Formula: C₂₀H₃₀Ni
MDL Number: MFCD00058704
CAS Number: 74507-63-4
UN: 1325
ADR: 4.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR