Bis(ethylcyclopentadienyl)nickel(II) ≥98%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Bis(ethylcyclopentadienyl)nickel

28-0083-1G 28-0083-5G
100205-064EA 172.68 USD
100205-064 100202-394
Bis(ethylcyclopentadienyl)nickel(II) ≥98%
Bis(ethylcyclopentadienyl)nickel (II)

Metallocenes, Derivatives & Cp Precursors, Volatile Organometallics for CVD & ALD, Volatile Precursors for CVD

Formula: C₁₄H₁₈Ni
MW: 244.986 g/mol
MDL Number: MFCD01862458
CAS Number: 31886-51-8
UN: 1993
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR