1,2-Bis(diphenylphosphino)ethane nickel(II) chloride ≥99%
Supplier: STREM CHEMICALS INC MS
Synonyms:
Dichloro[bis(1,2-diphenylphosphino)ethane]nickel(II)
28-0030-50G
28-0030-2G
28-0030-10G
100207-644EA
560.49
USD
100207-644
100201-564
100207-642
1,2-Bis(diphenylphosphino)ethane nickel(II) chloride ≥99%
1,2-Bis(diphenylphosphino)ethane nickel(II) chloride
Formula:
C₂₆H₂₄Cl₂NiP₂ MW: 528.016 g/mol |
MDL Number:
MFCD00013313 CAS Number: 14647-23-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...