Bis(1,5-cyclooctadiene)nickel(0) ≥98%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Bis(1,5-cyclooctadiene)nickel(0) (Wax encapsulated) (ca. 0.05mmol/capsule), Ni(COD)2

28-0010-10G 28-0010-2G
100198-168EA 496.8 USD
100198-168 100203-080
Bis(1,5-cyclooctadiene)nickel(0) ≥98%
Bis(1,5-cyclooctadiene)nickel (0)
Formula: C₁₆H₂₄Ni
MW: 275.06 g/mol
Melting Pt: 60 °C (N2) (dec.)
MDL Number: MFCD00058902
CAS Number: 1295-35-8
UN: 1325
ADR: 4.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR