Bis(pentamethylcyclopentadienyl)cobalt(III) hexafluorophosphate ≥98%

Supplier: STREM CHEMICALS INC MS

Synonyms: Bis(pentamethylcyclopentadienyl)cobalticinium hexafluorophosphate

27-1000-5G 27-1000-1G
100198-166EA 544.92 USD
100198-166 100197-472
Bis(pentamethylcyclopentadienyl)cobalt(III) hexafluorophosphate ≥98%
Bis(pentamethylcyclopentadienyl)cobalt(III) hexafluorophosphate

Metallocenes, Derivatives & Cp Precursors

Formula: C₂₀H₃₀CoF₆P
MW: 474.35 g/mol
MDL Number: MFCD00269818
CAS Number: 79973-42-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR