Cobalt(II) meso-tetraphenylporphine

Supplier: STREM CHEMICALS INC MS

Synonyms: Cobalt(II) TPP, Cobalt(II) Tetraphenylporphyrin, 5,10,15,20-Tetraphenyl-21H,23H-porphine Cobalt(II)

27-0470-5G 27-0470-1G
100204-606EA 986.52 USD
100204-606 100203-074
Cobalt(II) meso-tetraphenylporphine
Cobalt(II) meso-tetraphenylporphine

Porphine

Formula: C₄₄H₂₈CoN₄
MW: 671.65 g/mol
MDL Number: MFCD00010725
CAS Number: 14172-90-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR